Skip to Content
Merck
All Photos(1)

Key Documents

NIST2000

Calibration standard for high-resolution X-ray diffraction

NIST® SRM® 2000

Sign Into View Organizational & Contract Pricing

Select a Size

1 BLOCK
$5,020.00

$5,020.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 BLOCK
$5,020.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$5,020.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

solid

packaging

pkg of 1 block

manufacturer/tradename

NIST®

technique(s)

diffraction/scattering: suitable

application(s)

general analytical

General description

Each unit of Standard Reference Material (SRM) contains 25 mm × 25 mm × 0.725 mm double-polished (100)-oriented, single-crystal Si specimens with a nominal 50 nm Si0.85Ge0.15 epitaxial layer and 25 nm Si cap. For more information, please refer to the SDS and COA.


SRM 2000_cert

SRM 2000_SDS

Application

This Standard Reference Material (SRM) offers the high-resolution X-ray diffraction (HRXRD) community SI-traceable Si (220) d-spacing in transmission, wafer miscut angle relative to the crystal plane, and the surface-to-Si (004) Bragg angle in reflection for the specified reference wavelength.

Features and Benefits

Certified values for SRM 2000 are available and can be used to calibrate HRXRD instrumentation. Expiration details, along with instructions for use, are included on the NIST certificate.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service