Skip to Content
Merck
All Photos(1)

Key Documents

NIST1D

Limestone

NIST® SRM® 1d, argillaceous

Sign Into View Organizational & Contract Pricing

Select a Size

70 G
$2,640.00

$2,640.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
70 G
$2,640.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$2,640.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 70 g

manufacturer/tradename

NIST®

format

matrix material

SMILES string

[Ca+2].[O-]C(=O)[O-]

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in evaluating chemical and instrumental methods of analysis of limestone and related materials. SRM 1d is composed of argillaceous limestone quarried in Putnam County, Indiana, USA. A unit of SRM 1d consists of a single bottle containing approximately 70 g of material of which 97 % passes a 75 μm (No. 200) sieve. For more information, please refer to the COA and SDS.

SRM 1D_cert

SRM 1D_SDS

Other Notes

Certified for the analytes listed below.
Sodium oxide (Na2O), Magnesium oxide (MgO), Aluminium oxide (Al2O3), Silicon dioxide (SiO2), Phosphorus pentoxide (P2O5), Sulfur (S), Potassium oxide (K2O), Calcium oxide (CaO), Manganese (Mn), Iron(III) oxide (Fe2O3), Zinc oxide (ZnO), Strontium oxide (SrO)

Reference/Informational value is provided for the following analytes.
Carbon (C), Titanium dioxide (TiO2), Chromium(III) oxide (Cr2O3), Barium oxide (BaO), Beryllium (Be), Niobium (Nb), Cadmium (Cd), Neodymium (Nd), Cerium (Ce), Nickel (Ni), Chlorium (Cl), Praseodymium (Pr), Cesium (Cs), Rubidium (Rb), Dysprosium (Dy), Samarium (Sm), Erbium (Er), Tin (Sn), Europium (Eu), Terbium (Tb), Flurine (F), Thorium (Th), Gallium (Ga), Uranium (U), Gadolinium (Gd), Vanadium (V), Holmium (Ho), Yttrium (Y), Lanthanum (La), Ytterbium (Yb)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service