Skip to Content
Merck
All Photos(1)

Key Documents

NIST1993

Zeta Potential - Colloidal Silica (Nominal Mass Fraction 2.2 %)

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$862.00

$862.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
$862.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$862.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

packaging

pkg of 50 g

manufacturer/tradename

NIST®

application(s)

general analytical

format

single component solution

storage temp.

15-25°C

General description

Each unit of SRM contains two pre-scored ampoules, with each measuring ~25 mL of colloidal silica particles, suspended in an aqueous borate buffer with a pH of 8.9.
For more information, please refer to the SDS and COA.
SRM 1993_cert
SRM 1993_SDS

Application
SRM 1993 is intended primarily to evaluate the performance of instruments and techniques used to measure the zeta potential and electrophoretic mobility, particularly in electroacoustic methods requiring a high mass fraction and calibration.
It can also be used to calibrate electroacoustic instruments requiring relatively high mass fraction.

Features and Benefits

  • The NISTcertified values are determined using electrophoretic light scattering and valuesare traceable to the International System of Units (SI).
  • Provided with non-certifiedvalues ideal for use in method harmonization, methoddevelopment, and process control.
  • Expiration Certification, instructions for handling, storage,and uses are provided on the NIST certificate.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service