Skip to Content
Merck
All Photos(1)

Key Documents

NIST1949

Frozen Human Prenatal Serum

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

14.7 G
$3,070.00

$3,070.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
14.7 G
$3,070.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$3,070.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

form

liquid

packaging

pkg of 14.7456 g

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

application(s)

clinical testing

format

matrix material

storage temp.

-65 to -96°C

General description

A unit of SRM 1949 consists of two vials each of four levels, consisting of a base level (non-pregnant), and three pregnancy trimester levels (trimester 1, trimester 2, and trimester 3). Each vial contains approximately 1.8 mL of frozen human serum.

NIST1949_Cert

NIST1949_SDS

Application

This human prenatal serum standard reference material (SRM) is intended primarily for use in validating analytical methods for the determination of specified constituents in maternal human serum. This SRM can also be used for quality assurance when assigning values to in-house control materials.

Other Notes

  • Expiration details and storage and use instructions are provided on the NIST certificate
  • Notes on biomaterial handling, disposal, and transport documented in SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service