Skip to Content
Merck
All Photos(1)

Key Documents

NIST1945

Organics in whale blubber

NIST® SRM® 1945

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product NIST1945 is not currently sold in your country. Contact Technical Service

Quality Level

packaging

pkg of 2 x 15 g

manufacturer/tradename

NIST®

application(s)

food and beverages

General description

This Standard Reference Material (SRM) is a frozen whale blubber homogenate intended for use in evaluating analytical methods for the determination of selected polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, polybrominated diphenyl ether (PBDE) congeners, and toxaphene congeners. A unit of SRM 1945 consists of two screw-capped glass bottles, each containing approximately 15 g of frozen whale blubber homogenate. For more information, please refer to the COA and SDS.

SRM 1945_cert

SRM 1945_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service