Skip to Content
Merck
All Photos(1)

Documents

NIST1932

Fluorescein solution

NIST® SRM® 1932

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 3 x 2 mL

manufacturer/tradename

NIST®

application(s)

diagnostic assay manufacturing

format

single component solution

General description

This Standard Reference Material (SRM) is intended for use in establishing a reference scale for fluorescence intensity based upon Molecules of Equivalent Soluble Fluorophore (MESF) units. A unit of SRM 1932 consists of three flamed sealed amber glass ampoules, each containing approximately 2.0 mL of fluorescein in an aqueous borate buffer. For more information, plese refer to the SDS and COA.

SRM 1932_cert

SRM 1932_SDS

Other Notes

Certified for the analytes listed below.
Fluorescein

Reference/Informational value is provided for the following analytes.
Fluorescein, DHBBA, Ethanol, Methyl Isobutyl Ketone, Ethyl Acetate, Total Organics, Potassium (K), Sodium (Na), Chloride (Cl)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service