Skip to Content
Merck
All Photos(1)

Key Documents

NIST1693A

Sulfur Dioxide in Nitrogen (Nominal Amount-of-Substance Fraction 50 μmol/mol)

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

6 L
$9,070.00

$9,070.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
6 L
$9,070.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$9,070.00


Check Cart for Availability

Request a Bulk Order

form

gas

packaging

pkg of 8400 g (6 L cylinder)

manufacturer/tradename

NIST®

storage temp.

15-25°C

General description

This Standard Reference Material (SRM) is a primary gas mixture for which the amount-of-substance fraction, expressed as concentration, may be related to secondary working standards. This SRM is intended for the calibration of instruments used for sulfur dioxide determinations and for other applications.This SRM mixture is supplied in a DOT 3AL-specification aluminum (6061 alloy) cylinder with a water volume of 6 L.

SRM 1693A_cert

SRM 1693A_SDS

Other Notes

Certified for the analytes listed below.
Sulfur dioxide (SO2)

Reference/Informational value is provided for the following analytes.
Water (H2O), Total Hydrocarbons

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service