Skip to Content
Merck

Skip To

NIST1663A

Sulfur dioxide in nitrogen

NIST® SRM® 1663A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$14,230.00

About This Item

UNSPSC Code:
41116107

$14,230.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

packaging

pkg of cylinder

manufacturer/tradename

NIST®

format

mixture

General description

Other Notes

Certified for the analytes listed below.
Sulfur Dioxide (SO2)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
NIST1662ANIST1661ANIST1689
grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

-

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®

manufacturer/tradename

NIST®

format

mixture

format

mixture

format

mixture

format

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

packaging

pkg of cylinder

packaging

pkg of cylinder

packaging

pkg of cylinder

packaging

pkg of cylinder


Still not finding the right product?

Explore all of our products under Sulfur dioxide in nitrogen


pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Liquefied gas

Storage Class

2A - Gases

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters