Skip to Content
Merck
All Photos(1)

Key Documents

NIST1634C

Trace elements in fuel oil

NIST® SRM® 1634c

Synonym(s):

Fuel oil no. 6

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 100 mL

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

General description

Each unit of SRM 1634c contains 100 mL of fuel oil.

SRM 1634C_cert

SRM 1634C_SDS

Features and Benefits

  • Accompanied with certificate documenting NIST-certified mass fraction values for total elements in SRM1634C.
  • The reference and information values of elements are reported as mass fraction.
  • The certified values are metrological traceable to the SI unit of gram, expressed as milligrams per kilogram.
  • The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Cobalt (Co), Nickel (Ni), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Arsenic (As), Selenium (Se), Barium (Ba), Chlorine (Cl), Sodium (Na), Sulfur (S)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

141.8 °F

Flash Point(C)

61 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service