Skip to Content
Merck
All Photos(1)

Key Documents

NIST1549A

Whole milk powder

NIST® SRM® 1549a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product NIST1549A is not currently sold in your country. Contact Technical Service

grade

certified reference material

Quality Level

packaging

pkg of 5 pouches x 10 g ea

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A unit of SRM 1549a consists of five heat-sealed aluminized packets, each containing around 10 g of material. For more information, please refer to the SDS and COA.
SRM 1549A_cert
SRM 1549A_SDS

Application

The Standard Reference Material (SRM) is intended for use in the validation of methods for determining elements, vitamins, and cholesterol in whole milk powder and similar matrices and for quality assurance when assigning values to in-house control materials.

Features and Benefits

  • NIST certified values traceable to the International System of Units (SI) are derived unit of mass fraction, expressed as milligrams per kilogram or milligrams per gram.
  • Determination of elements was done using inductively coupled plasma optical emission spectrometry (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), and instrumental neutron activation analysis (INAA).
  • The determination of cholesterol was done by using isotope dilution (ID) gas chromatography (GC) with mass spectrometry (MS).
  • Expiration details, instructions for use, storage, and handling are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Ash, Carbohydrates, Fat (as the sum of fatty acids as Free Fatty acids), Phosphorous (P), Protein, Solids
Amino acids: Alanine, Arginine, Aspartic acid, Cysteine, Glutamic acid, Glycine, Histidine, Isoleucine, Leucine, Lysine, Methionine, Phenylalanine, Serine, Threonine, Tryptophan, Tyrosine, Valine
Lipids and Fatty acids: Butyric acid, Capric acid, Caproic acid, Caprylic acid, Cholesterol, DPA, Lauric acid, Linoleic acid, Margaric acid, Margaroleic acid, Myristic acid, Myristoleic acid, Oleic acid, Palmitic acid, Palmitoleic acid, Pentadecanoic acid (C15:0), Stearic acid, trans-Palmitelaidic acid, Tridecanoic acid (C13:0), Vaccenic acid, a-Linolenic acid
Trace Elements: Barium (Ba), Calcium (Ca), Copper (Cu), Iodine (I), Iron (Fe), Magnesium (Mg), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Potassium (K), Selenium (Se), Sodium (Na), Strontium (Sr), Zinc Zn)
Vitamins: 25-Hydroxyvitamin D3, Ascorbic acid (Vitamin C), Biotin, Carnitine, Cholecalciferol (Vitamin D3), Choline, Niacinamide (Vitamin B3), Pantothenic acid (Vitamin B5), Pyridoxal, Pyridoxamine (Vitamin B6), Riboflavin (Vitamin B2), Thiamine (Vitamin B1), Total Vitamin B6 as Pyridoxal, Vitamin B12

Matrix Group: Dairy products and eggs

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service