Skip to Content
Merck
All Photos(1)

Key Documents

NIST1247

Incoloy® 825

NIST® SRM® 1247

Sign Into View Organizational & Contract Pricing

Select a Size

1 DISC
$1,600.00

$1,600.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 DISC
$1,600.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,600.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

General description

Standard reference material Incoloy® 825 is a high-temperature alloy in wrought form. Each unit of SRM 1247 contains a single disk measuring around 35 mm in diameter and 19 mm in thickness. For more information, please refer to the COA and SDS.

SRM 1247_cert

SRM 1247_SDS

Application

SRM1247 is intended for use in validating chemical and instrumental analysismethods. It is also suitable for validating the value assignment of in-housereference materials.

Features and Benefits

  • Information mass fraction values for constituents are provided and expressed as Mass Fraction (mg/kg).
  • Expiration details, and use instructions are provided on the NIST certificate.
  • The mass fraction values for elements in nickel-base alloys are provided and estimated using a Gaussian random effects model and the DerSimonian-Laird procedure. These values are metrologically traceable to the SI derived unit for mass fraction, expressed as milligrams per kilogram.

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Carbon (C), Chromium (Cr), Cobalt (Co), Copper (Cu), Iron (Fe), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Niobium (Nb), Phosphorus (P), Silicon (Si), Titanium (Ti), Vanadium (V), Arsenic (As), Boron (B), Lead (Pb), Thallium (Tl)

Reference/Informational value is provided for the following analytes. Gallium (Ga), Nitrogen (N), Oxygen (O), Silver (Ag), Sulfur (S), Tungsten (W), Antimony (Sb), Tantalum (Ta), Bismuth (Bi), Tellurium (Te), Magnesium (Mg), Tin (Sn), Selenium (Se)

See certificate for values and more details at nist.gov/SRM.

Legal Information

Incoloy is a registered trademark of Special Metals Corporation
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service