Skip to Content
Merck
All Photos(1)

Key Documents

NIST1241C

Aluminum alloy 5182

NIST SRM 1241c

Sign Into View Organizational & Contract Pricing

Select a Size

1 DISC
$2,420.00

$2,420.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 DISC
$2,420.00

About This Item

UNSPSC Code:
12352311
NACRES:
NA.24

$2,420.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

One unit of SRM 1241C contains a single disk measuring ~ 6.3 cm in diameter and 1.9 cm in thickness.

SRM 1241C_cert

SRM 1241C_SDS

Application

Aluminum alloy 5182 SRM is intended for the evaluation of instrumental analysis methods including glow discharge optical emission spectrometry, spark source optical emission spectrometry, and X-ray fluorescence spectrometry.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified mass fraction values, expanded uncertainty, coverage factor for constituents in Aluminum alloy 5182.
  • The reference and information values of elements are reported as mass fraction.
  • The certified values are Metrologically traceable to the SI derived unit of mass, expressed as percent.
  • The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Silicon (Si)(b), Silicon (Si)(c), Iron (Fe), Copper (Cu), Manganese (Mn) Magnesium (Mg), Nickel (Ni), Zinc (Zn), Titanium (Ti)(b), Titanium (Ti)(c), Vanadium (V), Chromium (Cr), Gallium (Ga)

Reference/Informational value is provided for the following analytes.
Cadmium (Cd), Lead (Pb), Strontium (Sr), Tin (Sn), Zirconium (Zr)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service