Skip to Content
Merck
All Photos(1)

Key Documents

NA-BE2-1

nBer e 2 (Bertholletia excelsaallergen 2)

pkg of 250 μg

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$2,040.00

$2,040.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
250 μG
$2,040.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.21

$2,040.00


Estimated to ship on19 May 2025


Request a Bulk Order

grade

analytical standard

Assay

>95%

mol wt

(Multiple bands ranging from 12 to 50 kDa)

packaging

pkg of 250 μg

manufacturer/tradename

InBio

format

neat

storage temp.

−20°C

General description

Source: Brazil Nuts
Formulation: Preservative and carrier-free in 1X PBS, pH 7.4. Filtered through 0.22μm filter.
Concentration: See product insert.
Purification: Purified from Pichia pastoris culture by multi-step chromatography. Purity >95 % by SDS-PAGE

Notes:
(1) 11S globulins are synthesized as single peptide chain precursors that are cleaved post-translationally.
(2) Solubility of Ber e 2 is dependent on the presence of ?1M NaCl

Disclaimer

Allergens are provided for research and commercial use in vitro:  not for human in vivo or therapeutic use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service