Skip to Content
Merck
All Photos(1)

Key Documents

N5035

Sigma-Aldrich

Nalidixic acid

meets USP testing specifications

Synonym(s):

1,4-Dihydro-1-ethyl-7-methyl-1,8-naphthyridin-4-one-3-carboxylic acid, 1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O3
CAS Number:
Molecular Weight:
232.24
Beilstein:
750515
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Agency

USP/NF
meets USP testing specifications

form

powder

mp

227-229 °C (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

pharmaceutical (small molecule)

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

CCN1C=C(C(O)=O)C(=O)c2ccc(C)nc12

InChI

1S/C12H12N2O3/c1-3-14-6-9(12(16)17)10(15)8-5-4-7(2)13-11(8)14/h4-6H,3H2,1-2H3,(H,16,17)

InChI key

MHWLWQUZZRMNGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Biochem/physiol Actions

Used to modulate (inhibit) bacterial DNA gyrase-dependent processes such as DNA polymerization, (ATP-dependen)t DNA supercoiling, and chromosome fragmentation.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inna Chmelnitsky et al.
Antimicrobial agents and chemotherapy, 52(8), 2962-2965 (2008-06-04)
Fourteen out of 16 carbapenem-resistant quinolone-susceptible Enterobacter cloacae isolates were found to carry qnrB2 and bla(KPC-2) genes encoded on the same plasmid. One isolate also carried the aac(6')-Ib-cr gene. Coexistence of quinolone resistance determinants and bla(KPC-2) on the same plasmid
Valeria Bortolaia et al.
Antimicrobial agents and chemotherapy, 54(4), 1623-1626 (2010-01-27)
We characterized 67 Escherichia coli isolates with reduced susceptibility to cefotaxime or ceftiofur obtained from healthy broilers housed in five Italian farms. The bla(CTX-M-1), bla(CTX-M-32) and bla(SHV-12) beta-lactamase genes were identified on IncI1, IncN, or IncFIB plasmids. Considerable genetic diversity
Kate S Baker et al.
The Lancet. Infectious diseases, 15(8), 913-921 (2015-05-06)
Shigellosis is an acute, severe bacterial colitis that, in high-income countries, is typically associated with travel to high-risk regions (Africa, Asia, and Latin America). Since the 1970s, shigellosis has also been reported as a sexually transmitted infection in men who
Michelle D Brazas et al.
Antimicrobial agents and chemotherapy, 51(12), 4276-4283 (2007-09-26)
With few novel antimicrobials in the pharmaceutical pipeline, resistance to the current selection of antibiotics represents a significant therapeutic challenge. Microbial persistence in subinhibitory antibiotic environments has been proposed to contribute to the development of resistance. Pseudomonas aeruginosa cultures pretreated
Suzanne Bialek et al.
Antimicrobial agents and chemotherapy, 54(10), 4373-4378 (2010-08-04)
Cross-resistance to cefoxitin (FOX), chloramphenicol (CMP), and quinolones (nalidixic acid [NAL]) related to a putative efflux system overexpression has recently been reported for Klebsiella pneumoniae. The potential impact of this multidrug resistance (MDR) on the virulence of K. pneumoniae was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service