Skip to Content
Merck
All Photos(1)

Key Documents

N15405

Sigma-Aldrich

Nitrotetrazolium Blue chloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H30Cl2N10O6 · H2O · CH3OH
Molecular Weight:
867.69
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

impurities

<8 wt. % methanol

mp

189 °C (dec.) (lit.)

λmax

259 nm

SMILES string

[Cl-].[Cl-].CO.[H]O[H].COc1cc(ccc1-[n+]2nc(-c3ccccc3)n(n2)-c4ccc(cc4)[N+]([O-])=O)-c5ccc(c(OC)c5)-[n+]6nc(-c7ccccc7)n(n6)-c8ccc(cc8)[N+]([O-])=O

InChI

1S/C40H30N10O6.CH4O.2ClH.H2O/c1-55-37-25-29(13-23-35(37)47-41-39(27-9-5-3-6-10-27)45(43-47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-42-40(28-11-7-4-8-12-28)46(44-48)32-17-21-34(22-18-32)50(53)54;1-2;;;/h3-26H,1-2H3;2H,1H3;2*1H;1H2/q+2;;;;/p-2

InChI key

RAMYNPGXWWDCAB-UHFFFAOYSA-L

Application

Used in the detection of nucleic acid hybridization. Also used in the detection of ascorbate peroxidase activity in native gels by inhibition of reduction.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 2 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E P Diamandis et al.
Journal of clinical laboratory analysis, 7(3), 174-179 (1993-01-01)
We have used a double-stranded DNA probe linked to the cystic fibrosis locus to detect a single-copy gene from varying amounts of genomic DNA, with Southern blot analysis. The DNA plasmid probe was labeled with either biotin or digoxigenin. Biotin
R Mittler et al.
Analytical biochemistry, 212(2), 540-546 (1993-08-01)
A method for the detection of ascorbate peroxidase activity in native electrophoretic gels is described. The assay is based on the ability of ascorbate peroxidase to prevent the ascorbate-dependent reduction of nitroblue tetrazolium in the presence of H2O2. The method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service