Skip to Content
Merck
All Photos(1)

Key Documents

M2305005

Mitoxantrone impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Amino-5,8-dihydroxy-4-{{2-[(2-hydroxyethyl)amino]ethyl}amino}anthracene-9,10-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$221.00

$221.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$221.00

About This Item

Empirical Formula (Hill Notation):
C18H19N3O5
CAS Number:
Molecular Weight:
357.36
UNSPSC Code:
41116107
NACRES:
NA.24

$221.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

mitoxantrone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N(CCNc1c2c(c(cc1)N)C(=O)c3c(c(ccc3O)O)C2=O)CCO

InChI

1S/C18H19N3O5/c19-9-1-2-10(21-6-5-20-7-8-22)14-13(9)17(25)15-11(23)3-4-12(24)16(15)18(14)26/h1-4,20-24H,5-8,19H2

InChI key

NBSLQRRHRCDIHJ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mitoxantrone impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service