Skip to Content
Merck
All Photos(1)

Key Documents

M2280000

Minocycline hydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27N3O7 · HCl
CAS Number:
Molecular Weight:
493.94
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

pharmaceutical primary standard

API family

minocycline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2C[C@@H]3Cc4c(ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)N(C)C

InChI

1S/C23H27N3O7.ClH/c1-25(2)12-5-6-13(27)15-10(12)7-9-8-11-17(26(3)4)19(29)16(22(24)32)21(31)23(11,33)20(30)14(9)18(15)28;/h5-6,9,11,17,27,29-30,33H,7-8H2,1-4H3,(H2,24,32);1H/t9-,11-,17-,23-;/m0./s1

InChI key

GLMUAFMGXXHGLU-VQAITOIOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Minocycline hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maneesh Singh et al.
Immunology, 142(4), 562-572 (2014-01-15)
More than a quarter of a century of research has established chronic immune activation and dysfunctional T cells as central features of chronic HIV infection and subsequent immunodeficiency. Consequently, the search for a new immunomodulatory therapy that could reduce immune
M Ohhira et al.
Nihon Ronen Igakkai zasshi. Japanese journal of geriatrics, 32(1), 33-38 (1995-01-01)
This paper describes ultrasonically-guided infusion of minocycline hydrochloride solution (MINO infusion therapy) in benign nonparasitic cysts of the liver. MINO infusion therapy was performed in 7 large hepatic cysts and successful results were obtained in 6 lesions. The infusion procedure
Wen-tao Ni et al.
Pakistan journal of pharmaceutical sciences, 27(3), 463-467 (2014-05-09)
The shortage of effective antibiotics against multidrug-resistant Acinetobacter baumannii (MDR-Ab) has posed great threat to the public health. But the advent of tigecycline gives us new hope. The goal of our research was to assess the clinical efficacy of tigecycline
Black bones: minocycline-induced bone pigmentation.
Vishnusai Chauhan et al.
The Medical journal of Australia, 201(2), 114-114 (2014-07-22)
Thomas Schmitz et al.
Experimental neurology, 254, 153-165 (2014-02-05)
Poor neurological outcome in preterm infants is associated with periventricular white matter damage and hypomyelination, often caused by perinatal inflammation, hypoxia-ischemia, and hyperoxia. Minocycline has been demonstrated in animal models to protect the immature brain against inflammation and hypoxia-ischemia by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service