I3261
Indo 1-AM
93 - 100% purity
Synonym(s):
4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester
About This Item
Recommended Products
product name
Indo 1-AM, ~95% (HPLC)
Assay
~95% (HPLC)
Quality Level
fluorescence
λex 330 nm; λem 450 nm (with Calcium)
λex 356 nm; λem 478 nm in methanol
storage temp.
−20°C
SMILES string
CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)c1ccc(C)cc1OCCOc2cc(ccc2N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)-c3cc4ccc(cc4[nH]3)C(=O)OCOC(C)=O
InChI
1S/C47H51N3O22/c1-28-7-11-39(49(19-43(56)68-23-63-29(2)51)20-44(57)69-24-64-30(3)52)41(15-28)61-13-14-62-42-18-35(37-16-34-8-9-36(17-38(34)48-37)47(60)72-27-67-33(6)55)10-12-40(42)50(21-45(58)70-25-65-31(4)53)22-46(59)71-26-66-32(5)54/h7-12,15-18,48H,13-14,19-27H2,1-6H3
InChI key
CAWBRCOBJNWRLK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service