Skip to Content
Merck
All Photos(1)

Key Documents

H162

Supelco

m-Hydroxybenzoylecgonine

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$323.00

$323.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$323.00

About This Item

Empirical Formula (Hill Notation):
C16H19NO5
CAS Number:
Molecular Weight:
305.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$323.00


Estimated to ship on22 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

SMILES string

CN1[C@H]2CC[C@@H]1[C@H]([C@H](C2)OC(=O)c3cccc(O)c3)C(O)=O

InChI

1S/C16H19NO5/c1-17-10-5-6-12(17)14(15(19)20)13(8-10)22-16(21)9-3-2-4-11(18)7-9/h2-4,7,10,12-14,18H,5-6,8H2,1H3,(H,19,20)/t10-,12+,13-,14+/m0/s1

InChI key

XCJJCRPRQYXLRE-AHLTXXRQSA-N

General description

m-Hydroxybenzoylecgonine is a cocaine metabolite, which is commonly used as a potential biomarker for the determination of cocaine use.[1]

Application

m-Hydroxybenzoylecgonine may be used as an analytical reference standard for the quantification of the analyte in biological samples[2][3] using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous liquid chromatography-mass spectrometry quantification of urinary opiates, cocaine, and metabolites in opiate-dependent pregnant women in methadone-maintenance treatment
Shakleya.MD, et al.
Journal of Analytical Toxicology, 34(1), 17-25 (2010)
Development and validation of a liquid chromatography?atmospheric pressure chemical ionization-tandem mass spectrometry method for simultaneous analysis of 10 amphetamine-, methamphetamine-and 3, 4-methylenedioxymethamphetamine-related (MDMA) analytes in human meconium
Kelly T, et al.
Journal of Chromatography. B, Biomedical Applications, 867(2), 194-204 (2008)
S Pichini et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 820(1), 151-156 (2005-05-04)
A high-performance liquid chromatography (HPLC)-mass spectrometry (MS) assay, already validated for opiates and cocaine in meconium, has been re-applied for determination of m- and p-hydroxybenzoylecgonine, using nalorphine as the internal standard. Methodology included an initial extraction from the matrix by
L M Iwamoto et al.
Drug metabolism and disposition: the biological fate of chemicals, 28(3), 335-338 (2000-02-19)
Recently, meta-hydroxybenzoylecgonine (m-OH BE) was identified by gas chromatography-mass spectroscopy during quantitative analysis for cocaine. Identification of m-OH BE in addition to the routinely identified benzoylecgonine by gas chromatography-mass spectroscopy confirmatory assays may increase detection of cocaine-exposed infants and decrease
Paul P Robandt et al.
Journal of analytical toxicology, 32(8), 577-585 (2008-11-15)
A fully automated system utilizing a liquid handler and an online solid-phase extraction (SPE) device coupled with liquid chromatography-tandem mass spectrometry (LC-MS-MS) was designed to process, detect, and quantify benzoylecgonine (BZE), meta-hydroxybenzoylecgonine (m-OH BZE), para-hydroxybenzoylecgonine (p-OH BZE), and norbenzoylecgonine (nor-BZE)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service