Skip to Content
Merck
All Photos(1)

Key Documents

H1428000

Hydroxocobalamin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$313.00

$313.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
15 MG
$313.00

About This Item

Empirical Formula (Hill Notation):
C62H89CoN13O15P
CAS Number:
Molecular Weight:
1346.36
UNSPSC Code:
41116107
NACRES:
NA.24

$313.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

hydroxocobalamin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C62H89N13O14P.Co.H2O/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H2,63,77)(H2,64,78)(H2,65,79)(H2,66,80)(H2,67,81)(H2,68,82)(H,69,83)(H,85,86);;1H2/q-2;+2;-1/p-1/t31-,34+,35+,36+,37-,41+,52+,53+,56?,57-,59+,60-,61-,62-;;/m0../s1

InChI key

PJOHJFIDLYDPIZ-PMEYKKDOSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydroxocobalamin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Hydroxocobalamin].
Rumiko Kondo
Chudoku kenkyu : Chudoku Kenkyukai jun kikanshi = The Japanese journal of toxicology, 27(1), 59-62 (2014-04-15)
André L F Sampaio et al.
Mediators of inflammation, 2013, 741804-741804 (2013-06-20)
NOS/•NO inhibitors are potential therapeutics for sepsis, yet they increase clinical mortality. However, there has been no in vivo investigation of the (in vitro) •NO scavenger, cobalamin's (Cbl) endogenous effects on NOS/•NO/inflammatory mediators during the immune response to sepsis. We
[Advances in the clinical and laboratory studies on methylmalonic aciduria combined with homocysteinemia type cblC].
Yu-Peng Liu et al.
Zhonghua er ke za zhi = Chinese journal of pediatrics, 51(4), 313-316 (2013-08-10)
Transfusion Medicine Illustrated: an unusual cause of red plasma.
Eric Gehrie et al.
Transfusion, 54(1), 11-11 (2014-01-11)
Irene García et al.
Plant physiology, 162(4), 2015-2027 (2013-06-21)
Cyanide is produced concomitantly with ethylene biosynthesis. Arabidopsis (Arabidopsis thaliana) detoxifies cyanide primarily through the enzyme β-cyanoalanine synthase, mainly by the mitochondrial CYS-C1. CYS-C1 loss of function is not toxic for the plant and leads to an increased level of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service