Skip to Content
Merck
All Photos(1)

Key Documents

ERMDA470KIFCC

Human serum (serum proteins)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$391.00

$391.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$391.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$391.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

clinical testing

format

matrix material

storage temp.

−70°C

General description

ERM-DA470k/IFCC replaces the former product ERM-DA470. The new reference material is certified for 12 serum proteins. The material is the lyophilised form of processed serum, and is certified for A2M, AAG, AAT, ALB, C3C, C4, HPT, IgA, IgG, IgM, TRF and TTR.

Application

Human serum has been used in a study to assess the ability of a peptide-modified magnetic microbead for voltammetric sensing of soybean agglutinin (SBA) added in human serum.[1]

Analysis Note

For more information please see:
ERMDA470KIFCC

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuharu Sugawara et al.
Analytica chimica acta, 1001, 158-167 (2018-01-03)
For use in the voltammetric sensing of galactose-dependent proteins, we modified magnetic beads with a peptide that had both electroactive- and molecular recognition properties. The peptide consisted of a YXY sequence and behaved as an electron-transfer carbohydrate-mimetic peptide that would
Magnetic beads modified with an electron-transfer carbohydrate-mimetic peptide for sensing of a galactose-dependent protein
Sugawara K, et al.
Analytica Chimica Acta (2017)
Kazuharu Sugawara et al.
Analytica chimica acta, 1040, 166-176 (2018-10-18)
We used an electron-transfer carbohydrate-mimetic peptide (YYYYC) to construct an electrochemical cytosensing system. Magnetic beads were modified with either asialofetuin (ASF) or soybean agglutinin (SBA) to evaluate the effect on cell sensing. Because SBA binds to the galactose residue that
Cassandra L Crihfield et al.
Analytical chemistry, 93(3), 1537-1543 (2021-01-01)
Protein sieving, which is a fundamental tool in the biotechnology field, can be automated using capillary gel electrophoresis. The high-viscosity and biocompatible linear gels required for capillary sieving must be replaced for each run using high pressures. Thermally responsive gels

Questions

  1. What is the solution stability of Human serum (serum proteins) - ERMDA470KIFCC-1VL after reconstitution?

    1 answer
    1. If there is no microbial contamination during the reconstitution process, the solution of ERM-DA470k/IFCC can be used for one week. It has been verified that changes to the certified concentration observed during that period are not significant. Additional information can be found on Page 54. All documentation regarding the certification, stability, and storage of this product has been provided. Please refer to it for further details.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service