Skip to Content
Merck
All Photos(1)

Key Documents

ERMBF417A

Maize GMO Standard MON 863 x MON 810

for the detection of GMO Maize (MON 863 × MON 810) with PCR

Synonym(s):

GMO Maize Powder Standard 0% MON 863 x MON 810

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$208.00

$208.00


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
1 G
$208.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$208.00


Estimated to ship on28 March 2025


Request a Bulk Order

grade

certified reference material

product line

BioChemika

quality

for the detection of GMO Maize (MON 863 × MON 810) with PCR

composition

genetically modified MON 863 × MON 810 maize, 0%

manufacturer/tradename

JRC

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMBF417A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service