Skip to Content
Merck
All Photos(1)

Key Documents

D93503

Sigma-Aldrich

Silver diethyldithiocarbamate

ACS reagent, 99%

Synonym(s):

DETC, Diethyldithiocarbamic acid silver salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$142.00
25 G
$407.00

$142.00


Available to ship on10 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$142.00
25 G
$407.00

About This Item

Linear Formula:
(C2H5)2NCSSAg
CAS Number:
Molecular Weight:
256.14
Beilstein:
3718150
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

$142.00


Available to ship on10 April 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Assay

99%

form

powder

reaction suitability

core: silver
reagent type: catalyst

mp

172-175 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CCN(CC)C(=S)S[Ag]

InChI

1S/C5H11NS2.Ag/c1-3-6(4-2)5(7)8;/h3-4H2,1-2H3,(H,7,8);/q;+1/p-1

InChI key

NSVHDIYWJVLAGH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Silver diethyldithiocarbamate is used to introduce diethyldithiocarbamato ligand and for the determination of arsenic.[1]

Application

Also used for colorimetric determination of small quantities of arsenic
Reagent for the detection of arsenic[2][3][4] and for the introduction of the diethyldithiocarbamato ligand.[5]
Silver diethyldithiocarbamate (AgDDC) may be used in the preparation of adducts of triorganophosphines and diphosphines.[6]
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

Biochem/physiol Actions

Inhibits induction of macrophage nitric oxide synthase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and structural characterization of adducts of silver (I) diethyldithiocarbamate with P-donor ligands.
Di Nicola C, et al.
Inorgorganica Chimica Acta, 360(9), 2935-2943 (2007)
Inorganic Chemistry, 32, 3565-3565 (1993)
Hunjing Baohu, 23-23 (1991)
Evaluation of electrochemical hydride generation for spectrophotometric determination of As (III) by silver diethyldithiocarbamat
Arbab-Zavar MH and Hashemi M
Talanta, 52, 1007-1014 (2000)
Chem. Abstr., 117, 219549d-219549d (1992)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service