Skip to Content
Merck
All Photos(1)

Key Documents

D3060

Millipore

Dextrose starch agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171611
NACRES:
NA.74
Product D3060 is not currently sold in your country. Contact Technical Service

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

clinical testing
food and beverages

microbiology

suitability

nonselective for Neisseria spp.
nonselective for Streptococcus spp.

Application

Used to propagate Neisseria gonorrhoeae.

Components

Ingredients(g/L)
Proteose peptone15.00
Dextrose2.00
Starch, soluble10.00
Sodium chloride5.00
Disodium phosphate3.00
Gelatin20.00
Agar10.00

Preparation Note

Suspend 65 g of Dextrose Starch Agar in 1000 ml of distilled water. Boil to dissolve the medium completely. Dispense in tubes and sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Cool the tubed medium in a slanted position.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atlas, R.M.
Handbook of Microbiological Media (1993)

Articles

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service