Skip to Content
Merck
All Photos(1)

Key Documents

CRMUDELU

Density Standard

ASTM D 4052; ISO 12185 (0.8687 g/mL, 15 °C)

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$625.00

$625.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$625.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$625.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 250 mL

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Density Standards, ASTM D 4052 can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM), manufactured under UKAS accreditation

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service