Skip to Content
Merck
All Photos(1)

Documents

CRM42339

Supelco

Pyrethrum Extract Solution

certified reference material

Synonym(s):

Pyrethrum extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 1.2 mL

application(s)

cannabis testing

SMILES string

C\C(C)=C\C1[C@@H](C(=O)O[C@H]2CC(=O)C(C\C=C\C=C)=C2C)C1(C)C.COC(=O)\C(C)=C\C3[C@@H](C(=O)OC4CC(=O)C(C\C=C\C=C)=C4C)C3(C)C

InChI

1S/C22H28O5.C21H28O3/c1-7-8-9-10-15-14(3)18(12-17(15)23)27-21(25)19-16(22(19,4)5)11-13(2)20(24)26-6;1-7-8-9-10-15-14(4)18(12-17(15)22)24-20(23)19-16(11-13(2)3)21(19,5)6/h7-9,11,16,18-19H,1,10,12H2,2-6H3;7-9,11,16,18-19H,1,10,12H2,2-6H3/b9-8+,13-11+;9-8+/t16?,18?,19-;16?,18-,19-/m00/s1

InChI key

VXSIXFKKSNGRRO-YWUDCVDHSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Check out our entire range of pesticide mixes for Cannabis testing
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cannabis testing regulations include analysis of purity, potency, and contaminants. Learn more about analysis of cannabis for pesticides, mycotoxins, and pyrethrins registered in California by LC-MS/MS and GC-MS/MS Using Ascentis® Express Biphenyl and SLB®-5ms Columns

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service