Skip to Content
Merck
All Photos(1)

Key Documents

CRM013

Supelco

Trace Metals - Paint Chips

certified reference material, pkg of 50 g

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$1,440.00

$1,440.00


Available to ship on31 March 2025Details

This product incurs additional customs and shipping fees. For further information on price and availability, please contact your Account Manager, or the Technical Support Team on [email protected]

Request a Bulk Order

Select a Size

Change View
50 G
$1,440.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,440.00


Available to ship on31 March 2025Details

This product incurs additional customs and shipping fees. For further information on price and availability, please contact your Account Manager, or the Technical Support Team on [email protected]

Request a Bulk Order

grade

certified reference material

CofA

certificate is enclosed in each package

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM013-050

technique(s)

AAS: suitable
ICP: suitable

pH

6.35

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Cadmium
    Chromium
    Lead

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. what is the concentration range for this item CRM013-50g

    1 answer
    1. There are no specification ranges or concentration ranges for this product. Each lot will vary, and the certified values will be reported on the COA, an example of which can be found in the 'Documentation' section of the product page.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service