Skip to Content
Merck
All Photos(1)

Key Documents

CCW4105

Millipore

Plate Count Agar in tubes

Medium for aerobic plate counts by the surface inoculation method (standard Plate Count Agar) according to ISO 4833, 8552 & 17410 Standards, and IFU No. 6, suitable for microbiology

Synonym(s):

Aerobic Plate count Agar, Casein-Peptone dextrose Agar, PCA, Standard Methods Agar

Sign Into View Organizational & Contract Pricing

Select a Size

20 X 20 ML
$125.00

$125.00


Estimated to ship on19 May 2025


Request more information

Select a Size

Change View
20 X 20 ML
$125.00

About This Item

UNSPSC Code:
12352106
NACRES:
NA.74

$125.00


Estimated to ship on19 May 2025


Request more information

Agency

ISO 17410
ISO 4833
ISO 8552

Quality Level

form

solid

shelf life

12 mo. at 8‑25 °C

feature

ready-to-use

composition

agar, 15.00 g/L
casein peptone, 5.00 g/L
dextrose, 1.00 g/L
yeast extract, 2.50 g/L

packaging

box of 20 tubes

storage condition

(Tightly closed)

parameter

17 mL filling volume

tube volume

20 mL

color

yellowish

pH

7±0.2 (25 °C)

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

nonselective for

General description

The Plate Count Agar formulation is modified to avoid the addition of milk. The new composition allows the growth of most microorganisms without any further additions. The medium′s formulation is equivalent to that described by the ‘Standard Methods for the Examination of Dairy products’, the USP′s ‘Tryptone Glucose Yeast Agar’, the ‘Deutsche Landswirtchaft’ and the APHA, ISO, and AOAC′s Plate Count Agar. This is the medium of choice for the plate count of any type of sample.

Application

The Plate Count Agar is suitable for aerobic plate counts by the surface inoculation method (standard Plate Count Agar).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service