CCW4102
M-Chromogenic Coliform Broth in Tubes
Selective and differential medium for the detection and enumeration of coliforms and E.coli in water samples by MF technique, suitable for microbiology
Synonym(s):
M-CCB
Select a Size
1800 800 097.
This item is not able to be purchased as a guest user. For further information, please contact us atSelect a Size
About This Item
1800 800 097.
This item is not able to be purchased as a guest user. For further information, please contact us atRecommended Products
Quality Level
sterility
sterile
form
liquid
shelf life
9 mo. at 8‑25 °C
feature
ready-to-use
composition
chromogenic β GLU substrate, 0.20 g/L
chromogenic Salmon GAL subt, 0.20 g/L
disodium hydrogen phosphate, 2.70 g/L
peptone, 3.00 g/L
sodium chloride, 5.00 g/L
sodium dihydrogen phosphate, 2.20 g/L
sodium pyruvate, 1.00 g/L
sorbitol, 1.00 g/L
Tergitol™ 7, 0.15 g/L
L-tryptophan, 1.00 g/L
packaging
box of 50 tubes
storage condition
(Tightly closed)
parameter
2 mL filling volume
capacity
2 mL
tube volume
3 mL
color
pale yellow
pH
6.8±0.2 (25 °C)
application(s)
food and beverages
microbiology
suitability
Escherichia spp.
General description
Application
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service