Skip to Content
Merck
All Photos(2)

Key Documents

CAN1002

Supelco

Ethylene Oxide in DMA

certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1.2 ML
$82.30

$82.30


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1.2 ML
$82.30

About This Item

UNSPSC Code:
12352119
NACRES:
NA.21

$82.30


Estimated to ship on30 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 1.2 ML

storage temp.

2-30°C

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F

Flash Point(C)

64 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service