Skip to Content
Merck
All Photos(1)

Documents

C5901

Supelco

Codeine monohydrate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO3 · H2O
CAS Number:
Molecular Weight:
317.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

CN1[C@H](C2)[C@@]3([H])[C@@]4(CC1)C5=C2C=CC(OC)=C5O[C@@]4([H])[C@@H](O)C=C3.O

InChI

1S/C18H21NO3.H2O/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19;/h3-6,11-13,17,20H,7-9H2,1-2H3;1H2/t11-,12+,13-,17-,18-;/m0./s1

InChI key

WRRSFOZOETZUPG-FFHNEAJVSA-N

Application

Codeine monohydrate has been used as a standard in the quantitative analysis of codeine by using different types of protein assays like biuret, bicinchoninic acid (BCA), coomassie brilliant blue (CBB) , pyrogallol red–molybdate (PRM) assay, Lowry assay and benzethonium chloride (BEC) assay, while relating the effective response of the drug with the type of assay.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Weak narcotic analgesic, perhaps due to conversion to morphine, with minimal hypnotic properties; potent antitussive

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An evaluation of protein assays for quantitative determination of drugs.
Williams K M, et al.
Journal of Biochemical and Biophysical Methods, 57(1), 45-55 (2003)
K Krause et al.
Allergy, 68(2), 256-258 (2013-01-17)
Platelet-activating factor (PAF) causes wheal and flare responses which are abrogated by H1-antihistamines giving rise to the hypothesis that PAF-induced wheal development is secondary to histamine release from dermal mast cells. But is this hypothesis correct? Wheal and flare responses
Laurence Toms et al.
The Cochrane database of systematic reviews, (1)(1), CD001547-CD001547 (2009-01-23)
This is an updated version of the Cochrane review published in Issue 4, 1998. Combining drugs from different classes with different modes of action may offer opportunity to optimise efficacy and tolerability, using lower doses of each drug to achieve
Codeine phosphate in paediatric medicine.
D G Williams et al.
British journal of anaesthesia, 86(3), 413-421 (2001-09-28)
Is it farewell to codeine?
Brian J Anderson
Archives of disease in childhood, 98(12), 986-988 (2013-08-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service