Skip to Content
Merck
All Photos(1)

Key Documents

C4898

Supelco

Cottonseed flour

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$51.60

$51.60


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$51.60

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$51.60


Available to ship on03 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

format

neat

General description

Cottonseed flour is a high protein concentrate, which can be obtained from the kernel of the cottonseed after the removal of the oil from the seed.[1][2]

Application

A finely ground powder that gradually releases vitamins, minerals and proteins to microorganisms during fermentation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxicants Occurring Naturally in Foods, 624-624 (1973)
Technological Innovations in Major World Oil Crops, 405-405 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service