Skip to Content
Merck
All Photos(1)

Key Documents

C2360000

Clomipramine hydrochloride

for microbiological assay, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-Chloro-10,11-dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, Anafranil hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$254.00

$254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$254.00

About This Item

Empirical Formula (Hill Notation):
C19H23ClN2 · HCl
CAS Number:
Molecular Weight:
351.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$254.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

clomipramine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN(C)CCCN1c2ccccc2CCc3ccc(Cl)cc13

InChI

1S/C19H23ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-7,10-11,14H,5,8-9,12-13H2,1-2H3;1H

InChI key

WIMWMKZEIBHDTH-UHFFFAOYSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clomipramine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Central nervous system,Cardiovascular

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diagnosis and treatment of canine separation anxiety and the use of clomipramine hydrochloride (clomicalm).
D F Horwitz
Journal of the American Animal Hospital Association, 36(2), 107-109 (2000-03-24)
Ruyi Xu et al.
Journal of chromatography. A, 1350, 15-22 (2014-05-28)
A fast and efficient method for the determination of tricyclic antidepressants using electro-enhanced solid phase microextraction (SPME) coupled to gas chromatography-mass spectrometry (GC-MS) has been developed. One advantage of this approach is that the mass transfer of target analytes from
Carina Bäcktorp et al.
Journal of pharmaceutical sciences, 104(4), 1409-1420 (2015-02-26)
We have developed a predictive method, based on quantum chemical calculations, that qualitatively predicts N-oxidation by hydrogen peroxides in drug structures. The method uses linear correlations of two complementary approaches to estimate the activation barrier without calculating it explicitly. This
Knut Fredrik Seip et al.
Journal of chromatography. A, 1347, 1-7 (2014-05-06)
Electromembrane extraction (EME) was performed on samples containing substantial amounts of NaCl to investigate how the presence of salts affected the recovery, repeatability, and membrane current in the extraction system. A group of 17 non-polar basic drugs with various physical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service