Skip to Content
Merck
All Photos(1)

Key Documents

C203

Supelco

2-Chloro-2-deoxy-D-glucose

analytical standard

Synonym(s):

2-CIDG

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$625.00

$625.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
$625.00

About This Item

Empirical Formula (Hill Notation):
C6H11ClO5
CAS Number:
Molecular Weight:
198.60
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$625.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

form

solid

analyte chemical class(es)

oligosaccharides

storage condition

desiccated

technique(s)

GC/MS: suitable
HPLC: suitable
PET imaging: suitable
gas chromatography (GC): suitable

color

white

mp

139-145  °C

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](Cl)C=O

InChI

1S/C6H11ClO5/c7-3(1-8)5(11)6(12)4(10)2-9/h1,3-6,9-12H,2H2/t3-,4+,5+,6+/m0/s1

InChI key

RBEGMPAFDRYYIG-SLPGGIOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Hygroscopic. Air-sensitive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service