Skip to Content
Merck
All Photos(1)

Key Documents

BP531

(2-Butylbenzofuran-3-yl)(4-hydroxy-3,5-diiodophenyl)methanone

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

NSC 85437

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$321.00

$321.00


Estimated to ship on20 March 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$321.00

About This Item

Empirical Formula (Hill Notation):
C19H16I2O3
CAS Number:
Molecular Weight:
546.14
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$321.00


Estimated to ship on20 March 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

amiodarone

form

solid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Ic1c(c(cc(c1)C(=O)c2c3c([o]c2CCCC)cccc3)I)O

InChI

1S/C19H16I2O3/c1-2-3-7-16-17(12-6-4-5-8-15(12)24-16)18(22)11-9-13(20)19(23)14(21)10-11/h4-6,8-10,23H,2-3,7H2,1H3

InChI key

PNFMEGSMKIHDFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

(2-Butylbenzofuran-3-yl)(4-hydroxy-3,5-diiodophenyl)methanone BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeyu Chen et al.
Cell, 184(5), 1262-1280 (2021-02-27)
Improving effector activity of antigen-specific T cells is a major goal in cancer immunotherapy. Despite the identification of several effector T cell (TEFF)-driving transcription factors (TFs), the transcriptional coordination of TEFF biology remains poorly understood. We developed an in vivo T cell CRISPR screening
Hume Stroud et al.
Neuron, 107(5), 874-890 (2020-06-27)
The maturation of the mammalian brain occurs after birth, and this stage of neuronal development is frequently impaired in neurological disorders, such as autism and schizophrenia. However, the mechanisms that regulate postnatal brain maturation are poorly defined. By purifying neuronal
Mike R Wilson et al.
Cell reports, 33(6), 108366-108366 (2020-11-12)
Endometriosis affects 1 in 10 women and is characterized by the presence of abnormal endometrium at ectopic sites. ARID1A mutations are observed in deeply invasive forms of the disease, often correlating with malignancy. To identify epigenetic dependencies driving invasion, we
Miguel A Uc-Chuc et al.
International journal of molecular sciences, 21(13) (2020-07-09)
Despite the existence of considerable research on somatic embryogenesis (SE), the molecular mechanism that regulates the biosynthesis of auxins during the SE induction process remains unknown. Indole-3-acetic acid (IAA) is an auxin that is synthesized in plants through five pathways.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service