Skip to Content
Merck
All Photos(1)

Key Documents

BP284

Phenylephrine hydrochloride

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

(R)-(−)-Phenylephrine hydrochloride, (R)-(−)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, (R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$313.00

$313.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$313.00

About This Item

Linear Formula:
HOC6H4CH(CH2NHCH3)OH·HCl
CAS Number:
Molecular Weight:
203.67
Beilstein:
4158948
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$313.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

phenylephrine

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

143-145 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

OC1=CC([C@H](CNC)O)=CC=C1.Cl

InChI

1S/C9H13NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,9-12H,6H2,1H3;1H/t9-;/m0./s1

InChI key

OCYSGIYOVXAGKQ-FVGYRXGTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenylephrine hydrochloride BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Methylphenidate Prolonged-release Capsules
  • Methylphenidate Tablets
  • Phenylephrine Intracameral Injection
  • Phenylephrine Eye Drops
  • Phenylephrine Injection
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    related product

    Product No.
    Description
    Pricing

    Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Hazard Classifications

    Acute Tox. 4 Oral - Skin Sens. 1B

    Storage Class Code

    13 - Non Combustible Solids

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service