Skip to Content
Merck
All Photos(1)

Key Documents

BP253

Miconazole nitrate

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

(±)-Miconazole nitrate salt, 1-(2,4-Dichloro-β-[(2,4-dichlorobenzyl)oxy]phenethyl)imidazole

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$322.00

$322.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$322.00

About This Item

Empirical Formula (Hill Notation):
C18H14Cl4N2O · HNO3
CAS Number:
Molecular Weight:
479.14
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$322.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

miconazole

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

ClC1=CC(Cl)=CC=C1C(OCC2=CC=C(Cl)C=C2Cl)CN3C=CN=C3.[O-][N+](O)=O

InChI

1S/C18H14Cl4N2O.HNO3/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4)

InChI key

MCCACAIVAXEFAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Miconazole nitrate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Miconazole and Hydrocortisone Acetate Cream
  • Miconazole and Hydrocortisone Cream
  • Miconazole and Hydrocortisone Ointment
  • Miconazole Eye Drops
  • Miconazole Oromucosal Gel
  • Miconazole Cream
  • Biochem/physiol Actions

    Antifungal azole. Mode of action: Inhibits cytochrome P450-dependent 14α-demethylase, which is critical to ergosterol biosynthesis. The accumulated 14α-methylated sterols change the membrane structure of sensitive fungi, resulting in an altered cell membrane permeability. Also inhibits peroxidases, which results in accumulation of peroxide within the cell.
    Miconazole interacts with 14-α demethylase, a cytochrome P-450 enzyme necessary for ergosterol biosynthesis. The inhibition of ergosterol results in increased cellular permeability causing leakage of cellular contents. Miconazole may also inhibit endogenous respiration, interact with phospholipids in the cell membrane, inhibit the transformation of yeasts to mycelial forms, inhibit purine uptake, and interfere with triglyceride and phospholipid biosynthesis.

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Precautionary Statements

    Hazard Classifications

    Acute Tox. 4 Oral - Skin Sens. 1

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service