Skip to Content
Merck
All Photos(1)

Key Documents

BP218

Lymecycline

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

N6-[[[[(4S,4aS,5aS,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenyl]carbonyl]amino]methyl]-L-lysine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$372.00

$372.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$372.00

About This Item

Empirical Formula (Hill Notation):
C29H38N4O10
CAS Number:
Molecular Weight:
602.63
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$372.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

API family

lymecycline

form

solid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

InChI

1S/C29H38N4O10/c1-28(42)13-7-6-9-17(34)18(13)22(35)19-14(28)11-15-21(33(2)3)23(36)20(25(38)29(15,43)24(19)37)26(39)32-12-31-16(27(40)41)8-4-5-10-30/h6-7,9,14-16,21,31-32,34-35,39,42-43H,4-5,8,10-12,30H2,1-3H3,(H,40,41)/b26-20-/t14?,15-,16?,21?,28+,29-/m0/s1

InChI key

NKQBBDWEXUNSIS-IOAYUFSMSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lymecycline BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Lymecycline Capsules
  • Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Yang Chen et al.
    Pharmacological research, 159, 105007-105007 (2020-06-21)
    Epidermal growth factor receptor-tyrosine kinase inhibitors (EGFR-TKIs) were first-line treatments for NSCLC patients with EGFR-mutations. However, about 30 % of responders relapsed within six months because of acquired resistance. In this study, we used Connectivity Map (CMap) to discover a
    Asha Ashuo et al.
    Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 37(9), 1481-1490 (2020-07-28)
    Herein, we have successfully established a novel, rapid, and simple lateral-flow immunoassay based on time-resolved fluorescence and biotin-streptavidin to detect the residues of various antibiotics in milk. The fluorescence signal and sensitivity of immunochromatography were enhanced through biotinylated antibody coupled

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service