Skip to Content
Merck
All Photos(1)

Key Documents

BP143

Ethacrynic acid

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

(2,3-Dichloro-4-[2-methylenebutyryl]phenoxy)acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$379.00

$379.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
100 MG
$379.00

About This Item

Empirical Formula (Hill Notation):
C13H12Cl2O4
CAS Number:
Molecular Weight:
303.14
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$379.00


0 left in stock

Request a Bulk Order

grade

pharmaceutical primary standard

API family

ethacrynic acid

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C(C(C(CC)=C)=O)=CC=C1OCC(O)=O

InChI

1S/C13H12Cl2O4/c1-3-7(2)13(18)8-4-5-9(12(15)11(8)14)19-6-10(16)17/h4-5H,2-3,6H2,1H3,(H,16,17)

InChI key

AVOLMBLBETYQHX-UHFFFAOYSA-N

Gene Information

human ... SLC12A1(6557)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethacrynic acid BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service