Skip to Content
Merck
All Photos(1)

Key Documents

BCR682

Mussel tissue (PCB′s)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

70 G
$744.00

$744.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
70 G
$744.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$744.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

matrix

mussel

format

matrix material

storage temp.

2-8°C

Analysis Note

For more information please see:
BCR682

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants: PCB 28, PCB 52, PCB 118, PCB 138, PCB 149, PCB 153, PCB 170, PCB 180

Matrix Group: Fish and Seafood

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service