Skip to Content
Merck
All Photos(1)

Key Documents

BCR663

Saxitoxin in acetic acid (determination in mussel)

BCR®, certified reference material

Synonym(s):

Saxitoxin

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$388.00

$388.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$388.00

About This Item

Beilstein:
47858
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

$388.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

application(s)

pharmaceutical (small molecule)

format

matrix material

SMILES string

[H][C@@]12N=C(N)N[C@]13N(CCC3(O)O)C(N)=N[C@H]2COC(N)=O

InChI

1S/C10H17N7O4/c11-6-15-5-4(3-21-8(13)18)14-7(12)17-2-1-9(19,20)10(5,17)16-6/h4-5,19-20H,1-3H2,(H2,12,14)(H2,13,18)(H3,11,15,16)/t4-,5-,10-/m0/s1

InChI key

RPQXVSUAYFXFJA-HGRQIUPRSA-N

Analysis Note

For more information please see:
BCR663

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service