BCR486
Purified human alfafoetoprotein (protein mass)
BCR®, certified reference material
Synonym(s):
Human alfafoetoprotein
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
certified reference material
Agency
BCR®
manufacturer/tradename
JRC
application(s)
clinical
format
matrix material
storage temp.
−20°C
Analysis Note
For more information please see:
BCR486
BCR486
Legal Information
BCR is a registered trademark of European Commission
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service