Skip to Content
Merck
All Photos(1)

Key Documents

BCR156R

Dibenz[c,h]acridine

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$321.00

$321.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$321.00

About This Item

Empirical Formula (Hill Notation):
C21H13N
CAS Number:
Molecular Weight:
279.33
Beilstein:
209259
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$321.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)ccc3cc4ccc5ccccc5c4nc23

InChI

1S/C21H13N/c1-3-7-18-14(5-1)9-11-16-13-17-12-10-15-6-2-4-8-19(15)21(17)22-20(16)18/h1-13H

InChI key

BTVBCAKHMZHLFR-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR156R

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Myriam Coulet et al.
Toxicology in vitro : an international journal published in association with BIBRA, 59, 281-291 (2019-05-06)
Nitrogen-containing polycyclic aromatic hydrocarbons (PANHs or azaarenes) are compounds structurally similar to PAHs (carbon substituted by a nitrogen) reported to occur at low levels in food. Although limited, literature may suggest possible higher toxicity than for PAHs. Using a battery
A H Conney et al.
Advances in experimental medicine and biology, 500, 697-707 (2002-01-05)
Chinese hamster V79 cells were exposed to a high or low concentration of the highly carcinogenic (R,S,S,R) or the less active (S,R,R,S) bay- or fjord-region diol epoxides of benzo[a]pyrene, benzo[c]phenanthrene or dibenz[c,h]acridine. Independent 8-azaguanine-resistant clones were isolated, and base substitutions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service