Skip to Content
Merck
All Photos(1)

Key Documents

BCR137R

Benzo[b]naphtho[1,2-d]thiophene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$321.00

$321.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$321.00

About This Item

Empirical Formula (Hill Notation):
C16H10S
CAS Number:
Molecular Weight:
234.32
Beilstein:
9635
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$321.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)ccc3sc4ccccc4c23

InChI

1S/C16H10S/c1-2-6-12-11(5-1)9-10-15-16(12)13-7-3-4-8-14(13)17-15/h1-10H

InChI key

XZUMOEVHCZXMTR-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR137R

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose Juan Rodríguez et al.
Organic & biomolecular chemistry, 10(36), 7334-7346 (2012-08-01)
Based on the benzo[b]naphtho[1,2-d]furan and benzo[b]naphtho[1,2-d]thiophene frameworks, a series of ligands with different basic side chains (BSCs) has been synthesized and pharmacologically evaluated. Also, their binding modes have been modelled using docking techniques. It was found that the introduction of
J Jacob et al.
Cancer letters, 32(1), 107-116 (1986-07-01)
Thiaarenes are metabolized by liver microsomes of untreated rats predominantly to sulfones and sulfoxides. After pretreatment of rats with monooxygenase inducers, ring oxidation of thiaarenes is also observed. In case of benzo[b]naphtho[2,3-d]thiophene the formation of a p-quinone takes place. Rat

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service