Skip to Content
Merck
All Photos(3)

Key Documents

B3022

Supelco

Beclomethasone dipropionate

analytical standard, for drug analysis

Synonym(s):

9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$136.00
1 G
$356.00
5 G
$1,380.00

$136.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$136.00
1 G
$356.00
5 G
$1,380.00

About This Item

Empirical Formula (Hill Notation):
C28H37ClO7
CAS Number:
Molecular Weight:
521.04
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$136.00


Estimated to ship on31 May 2025


Request a Bulk Order

Quality Level

Assay

≥99%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C

InChI

1S/C28H37ClO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1

InChI key

KUVIULQEHSCUHY-XYWKZLDCSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Beclomethasone dipropionate belongs to the chemical class of chlorinated glucocorticoid diesters, well known for its use in the treatment of asthma due to its anti-inflammatory properties.

Application

Beclomethasone dipropionate analytical standard can be used as follows:
  • Development of a reversed-phase high-performance liquid chromatographic (RP-HPLC) method, combined with UV detection to determine beclomethasone dipropionate in nanocapsule suspensions
  • Simultaneous determination of beclomethasone dipropionate and ketoconazole in their bulk and combined cream formulations using UV-spectrophotometry and RP-HPLC based methods
  • Estimation of beclomethasone dipropionate and formoterol fumarate in their combined dosage forms by stability indicating RP-HPLC method coupled with UV detection
  • Combined analysis of beclomethasone dipropionate and formoterol fumarate dihydrate using two UV-spectrometric and one RP-HPLC based methods in bulk and their pharmaceutical dosage forms
  • Multi-residue analysis of 21 glucocorticoids in environmental samples by ultra-high performance liquid chromatography (UHPLC) and mass spectrometry (MS)

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Adrenal gland,Immune system,Bone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quick Review of Pharmacology null
Linjie Zhang et al.
Respirology (Carlton, Vic.), 18(2), 272-277 (2012-10-09)
Recent studies have raised concerns about the link between use of inhaled corticosteroids (ICS) and risk of pneumonia in patients with chronic obstructive pulmonary disease. This cross-sectional study aimed to investigate the association between ICS and oropharyngeal colonization by Streptococcus
Eli O Meltzer et al.
Allergy and asthma proceedings, 33(3), 249-257 (2012-06-28)
Intranasal corticosteroids are recommended as first-line therapy for the treatment of the symptoms of persistent allergic rhinitis (AR). Since the phase-out of chlorofluorocarbon nasal aerosols, intranasal corticosteroids have been available only as aqueous nasal sprays. This study was designed to
Kai-Michael Beeh et al.
The Journal of allergy and clinical immunology, 131(3), 866-874 (2013-02-07)
Allergen-specific TH2 responses contribute to the development of allergic asthma. Their increase may be due to a reduced early exposure to environmental pathogens, which induces a TH1 response, and thereby suppresses the allergic TH2 response. QbG10 (bacteriophage Qbeta-derived virus-like particle
William J Anderson et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 109(3), 185-189 (2012-08-28)
Whether small airways dysfunction persists in patients with asthma receiving standard community treatment is unknown. Impulse oscillometry (IOS) is a sensitive measure of small airways function. To assess the degree of small airways dysfunction in a cross-section of patients with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service