Skip to Content
Merck
All Photos(1)

Key Documents

B0070000

Bacampicillin hydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N3O7S · HCl
CAS Number:
Molecular Weight:
501.98
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bacampicillin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

S1[C@H]2N([C@H](C1(C)C)C(=O)OC(OC(=O)OCC)C)C(=O)[C@H]2NC(=O)[C@H](N)c3ccccc3.[Cl-].[H+]

InChI

1S/C21H27N3O7S.ClH/c1-5-29-20(28)31-11(2)30-19(27)15-21(3,4)32-18-14(17(26)24(15)18)23-16(25)13(22)12-9-7-6-8-10-12;/h6-11,13-15,18H,5,22H2,1-4H3,(H,23,25);1H/t11?,13-,14-,15+,18-;/m1./s1

InChI key

IWVTXAGTHUECPN-ANBBSHPLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bacampicillin hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of two bacampicillin therapeutic regimens in infections of middle ear, pharynx and paranasal sinuses.
A Ottaviani et al.
Journal of chemotherapy (Florence, Italy), 1(4 Suppl), 750-751 (1989-07-01)
Y Akimoto et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 50(1), 11-13 (1992-01-01)
Ampicillin concentrations in human serum and radicular granulomas of 42 patients were determined after a single oral dose of bacampicillin (equivalent to 500 mg of ampicillin). Although wide variations were found among both serum and radicular granuloma ampicillin concentrations, measurable
G Reid et al.
Scandinavian journal of infectious diseases, 22(1), 43-47 (1990-01-01)
Specimens were examined from 70 female presenting with urinary tract infection. 50 patients were randomly treated with 7 days amoxicillin or bacampicillin and another 20 randomly received amoxicillin or enoxacin. Effective clearance of the infections was achieved with each antibiotic
M Bogataj et al.
Journal of microencapsulation, 8(3), 401-406 (1991-07-01)
Eudragit E microspheres containing bacampicillin hydrochloride were prepared by solvent evaporation and solvent extraction methods. Three different systems of solvents were used: methyl acetate, acetone and methanol/liquid paraffin. The success of the procedures depended mostly on the lipophilicity of the
A comparative pharmacokinetic study between lenampicillin, bacampicillin and amoxycillin.
A M Sefton et al.
Journal of chemotherapy (Florence, Italy), 1(4 Suppl), 494-495 (1989-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service