Skip to Content
Merck
All Photos(1)

Key Documents

ATG002

Sigma-Aldrich

A549 EGFR Antigen Panel Cell Lines

Synonym(s):

A549 Cell Panel, A549 EGFR Cell Lines, EGFR Antigen Panel

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$19,140.00

$19,140.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
$19,140.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$19,140.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

human lung (carcinoma)

growth mode

adherent

morphology

epithelial

shipped in

dry ice

storage temp.

−196°C

General description

This product is a panel of three (3) genetically modified cell lines involving the EGFR gene. The panel consists of a EGFR knockout cell line (EGFR-/-) as well as a EGFRlow and a EGFRhigh expression cell line. The EGFR-/- cell line was generated using CompoZr® Zinc Finger Nuclease technology, and the two EGFR expression cell lines were generated using MISSION® lentivirus. The EGFRlow and EGFRhigh cell lines express EGFR at low and high levels relative to the EGFR-/- cell line (EGFRhigh > EGFRlow > EGFR-/-) as measured via fluorescence-activated cell sorting (FACS). As a comparison, wild type A549 cells demonstrate EGFRmedium expression as measured via FACS.

The STR profile of these cell lines match that of its parental cell line - ECACC catalog number 86012804.

Cell Line Description

Derived from a 58 year old Caucasian male. The cells can synthesise lecithin utilising the cytidine diphosphocholine pathway. Occasional cells may also contain inclusion bodies although they are not known to carry any human pathogen.

Features and Benefits

The A549 EGFR antigen panel cell lines feature EGFR expressed at varying levels (KO, low, and high). These cell lines enable researchers to screen and evaluate the efficacy and potency of EGFR-targeted immunotherapies in a biologically-relevant cellular research model.

Components

ATG002A-1VL: A549 EGFR Knockout
ATG002B-1VL: A549 EGFR Low
ATG002C-1VL: A549 EGFR High

Quality

Tested for Mycoplasma, bacterial and fungal content, post-freeze viability, and short terminal repeat (STR) analysis for cell line identification.

Legal Information

CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service