A4045
Alcian Blue-tetrakis(methylpyridinium) chloride
Synonym(s):
A,B,C,D-tetrakis(Pyridiniomethyl) Cu(II) phthalocyanine chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C56H40Cl4CuN12
CAS Number:
Molecular Weight:
1086.36
UNSPSC Code:
12171500
Recommended Products
composition
Dye content, ~90%
SMILES string
[Cl-].[Cl-].[Cl-].[Cl-].C(c1cccc2c3nc(nc4n5[Cu]n6c(nc7nc(nc5c8c(C[n+]9ccccc9)cccc48)c%10cccc(C[n+]%11ccccc%11)c7%10)c%12cccc(C[n+]%13ccccc%13)c%12c6n3)c12)[n+]%14ccccc%14
Linkage
This dye is similar in structure and properties to Alcian Blue 8GX, with pyridiniomethyl substituents in each ring instead of amidinothiomethyl groups.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Atukorallaya Devi Sewvandini Atukorala et al.
Developmental biology, 441(2), 235-241 (2018-07-19)
The phenotype of lens-ablated Mexican tetra (Astyanax mexicanus) compared to wild-type surface fish has been described and includes, among other effects, eye degeneration, changes in tooth number and cranial bone changes. Here, we investigate the spatiotemporal expression patterns of several
Florian Ponzio et al.
Journal of colloid and interface science, 459, 29-35 (2015-08-12)
Polydopamine (PDA) coatings appear as a universal functionalization methodology allowing to coat the surface of almost all kinds of known materials with a conformal, stable, robust and reactive material. Relatively few investigations were dedicated to the incorporation of other molecules
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service