Skip to Content
Merck
All Photos(2)

Key Documents

A3299

Sigma-Aldrich

Bovine Serum Albumin solution

30% in 0.85% sodium chloride, aseptically filled

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$227.00
250 ML
$855.00
1 L
$2,620.00

$227.00


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 ML
$227.00
250 ML
$855.00
1 L
$2,620.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

$227.00


Available to ship on07 April 2025Details


Request a Bulk Order

biological source

bovine

Quality Level

sterility

aseptically filled

Assay

28-32% (biuret)

form

solution

quality

high avidity

purified by

heat shock fractionation

contains

0.1% sodium azide as preservative

does not contain

stabilizer

origin

USA origin

concentration

28-32% (biuret)
30% in 0.85% sodium chloride

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

General description

Bovine serum albumin (BSA) is a member of the serum albumin family.[1] It is a globular, α-helical, and non-glycosylated protein.[2] BSA is a single-chain protein and consists of three domains with two sub-domains under each.[2] It has 17-disulfide bonds.[1]

Application

Bovine Serum Albumin solution has been used as a standard for the quantification of protein in extracellular polymeric substances (EPSs) using the Lowry method.[3]

Biochem/physiol Actions

Bovine Serum Albumin (BSA) facilitates the transport of drugs, hormones, and fatty acids.[2][4] It is used as a blocking agent in enzyme-linked immunosorbent assay (ELISA).[5] BSA is an important constituent of the cell culture media and favors embryonic stem cells (hESC) differentiation.[6] It plays a vital role in the cosmetic and pharmaceutical industry.[4]

Quality

High avidity

Analysis Note

Tested for avidity with known incomplete antibody (not saline agglutinable).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasound, thermal and alkali treatments affect extracellular polymeric substances (EPSs) and improve waste activated sludge dewatering
Ruiz-Hernando M, et al.
Process Biochemistry (Oxford, United Kingdom), 50(3), 438-446 (2015)
Subhajit Ghosh et al.
The journal of physical chemistry. B, 119(25), 7804-7815 (2015-05-30)
The study of protein-surfactant interactions is important because of the widespread use of surfactants in industry, medicine, and pharmaceutical fields. Sodium N-lauroylsarcosinate (SL-Sar) is a widely used surfactant in cosmetics, shampoos. In this paper, we studied the interactions of bovine
Rebeka Popovic et al.
Disease models & mechanisms, 16(4) (2023-04-21)
Eukaryotic Tribbles proteins are pseudoenzymes that regulate multiple aspects of intracellular signalling. Both Drosophila melanogaster and mammalian members of this family of pseudokinases act as negative regulators of insulin signalling. Mammalian tribbles pseudokinase (TRIB) genes have also been linked to
Structural changes during the unfolding of Bovine serum albumin in the presence of urea: A small-angle neutron scattering study
Das A, et al.
Pramana, 63(2), 363-368 (2004)
Yuhong Xiao et al.
Journal of immunological methods, 384(1-2), 148-151 (2012-06-27)
The enzyme-linked immunosorbent assay (ELISA) is an extremely common and powerful laboratory technique for detecting proteins by antibodies. Researchers frequently use bovine serum albumin (BSA) as a blocking agent to prevent non-specific binding of antigens and antibodies to the microtiter

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service