Skip to Content
Merck
All Photos(1)

Key Documents

A-2045-040

TOC-Calibration Kit

25 mg TOC

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$717.00

$717.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$717.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.21

$717.00


Check Cart for Availability

Request a Bulk Order

manufacturer/tradename

Altus

technique(s)

TOC: suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

single component solution

Looking for similar products? Visit Product Comparison Guide

General description

Total organic carbon (TOC) is a measure of the total amount of carbon in an organic compound. It is a critical parameter to evaluate the water quality for possible organic contamination

Application

This kit is intended to be used for the calibration and validation of analytical instruments during the measurement of total organic carbon (TOC) for drinking water, groundwater, surface water, seawater, and wastewater.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service